Learn More
DWK Life Sciences Wheaton™ Socorex Smarties Caps For Socorex Acura Manual Pipettes
Shop All DWK Life Sciences Products

Description
- Pick among 14 colors or choose the smartie mix for other color combinations
Specifications
Specifications
| Color | Green |
| Material | Plastic |
| Type | Smarties Caps For Socorex Acura Pipette |
| For Use With (Equipment) | Socorex™ Acura™ manual pipette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.