Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Socorex Smarties Caps For Socorex Acura Manual Pipettes
SDP

Catalog No. 1370970 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Assorted
Blue
Gray
Green
Ice Blue
Lemon
Mint
Orange
Pink
Purple
Red
Rose
Vanilla
White
Yellow
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
13-709-70 Assorted
13-709-56 White
13-709-64 Pink
13-709-58 Vanilla
13-709-61 Orange
13-709-65 Purple
13-709-67 Blue
13-709-57 Gray
13-709-59 Lemon
13-709-60 Yellow
13-709-62 Red
13-709-63 Rose
13-709-66 Ice Blue
13-709-68 Mint
13-709-69 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 13-709-70 Supplier DWK Life Sciences Supplier No. W820014
Add to Cart
Add to Cart

Color coding for easy identification - Acura™ pipettes color-coded button caps for individual, lab or department identifications.

  • Pick among 14 colors or choose the smartie mix for other color combinations
TRUSTED_SUSTAINABILITY

Specifications

Color Assorted
Material Plastic
Type Smarties Caps For Socorex Acura Pipette
For Use With (Equipment) Socorex™ Acura™ manual pipette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.