Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Socorex Calibra™ 822 Digital Micropipette from DWK Life Sciences™
SDP

Catalog No. 137071 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
0.2 to 2 μL
1 to 10 μL
2 to 20 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-707-1 0.2 to 2 μL
13-707-2 1 to 10 μL
13-707-2A 2 to 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-707-1 Supplier Socorex Supplier No. 851160

For gel sequencing and biotechnology applications

  • Single-channel, variable volume with twin cams
  • Precalibrated steps to provide true digital volume entry
  • Color-coded to ensure proper tip selection

TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±6%, <±4%, <±2%
Certifications/Compliance CE, IVD 98/79 EEC, GMP, GLP, NCCLS
Compatible Tips Ultra 10μL tip
Autoclavable Autoclavable
For Use With Ultra 10 μL Tip
Model Digital 822
Number of Channels 1
Product Line Socorex Calibra
Type Micropipette
Volume (Metric) 0.2 to 2 μL
For Use With (Application) Gel sequencing and biotechnology applications
For Use With (Equipment) Ultra 10μL tip
Increments 0.1μL
UV Monitoring Yes
Warranty 3 Years
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.