Learn More
DWK Life Sciences Wheaton™ Square Media Bottles with Tamper Evident Caps
Shop All DWK Life Sciences Products
Description
- For storage of media, laboratory reagents and biological buffers
- Made of Eastman Tritan Eastar Copolyester PETG that is resin used for single use labware and medical devices
- With tamper evident blue polypropylene caps
- Molded graduation markings for reference filling
- Gamma irradiated sterilized to SAL 10-6
- Cleanroom packaged for sterile production with multi-barrier elastomer packaging
- Resin meets ISO 10993 and/or USP Class VI biocompatibility requirements and is Food Contact Status compliant
Specifications
Specifications
| Capacity (Metric) | 125 mL |
| Material | PETG |
| Sterility | Sterile |
| Shape | Square |