Learn More
DWK Life Sciences Stainless Steel Microtest Manifold
Make dispensing easier and quicker by selecting a Stainless Steel manifold dispensing system.
$594.00 - $912.00
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Material | Stainless Steel |
| Product Type | Microtest Manifold |
| For Use With (Equipment) | Microplates |
| Catalog Number | Mfr. No. | Needle Point Number | Needle Point Style | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Needle Point Number | Needle Point Style | Price | Quantity & Availability | ||||
02-912-802
![]() |
DWK Life Sciences
851388 |
12 Channels | Straight |
Case for $912.00
|
|
||||
02-912-801
![]() |
DWK Life Sciences
851382 |
4 Channels | Angled |
Case for $594.00
|
|
||||
02-912-803
![]() |
DWK Life Sciences
851389 |
12 Channels | Angled |
Case for $912.00
|
|
||||
Description
- Unique dispensing system enables performance of sample inoculations and cell feedings faster and more efficiently under sterile conditions
- Ideal for use with 24 and 96 well microplates
Specifications
| Autoclavable | |
| Stainless Steel | |
| Microtest Manifold | |
| Microplates |