Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Straight Plug Rubber Stoppers
SDP

Catalog No. 0645150 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Black
Gray
Red
Diameter (Metric) Outer:
13 mm
20 mm
28 mm
Material:
Chlorobutyl-Isoprene
FEP
Rubber
Viton
Product Type:
Straight Plug Stopper
Two-leg Lyophilization Stopper
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Diameter (Metric) Outer Material Product Type
06-451-50 Red 20 mm Rubber Straight Plug Stopper
06-406-11A Red 13 mm Rubber Straight Plug Stopper
06-451-53 Gray 20 mm FEP Straight Plug Stopper
06-451-52 Gray 20 mm Chlorobutyl-Isoprene Straight Plug Stopper
06-449-13 Gray 28 mm Chlorobutyl-Isoprene Two-leg Lyophilization Stopper
06-451-54 Black 20 mm Viton Straight Plug Stopper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 06-451-50 Supplier DWK Life Sciences Supplier No. 224100170

Provides maximum sealing for vials with straight-wall glass finishes. Wheaton™ Straight Plug Rubber Stoppers are manufactured from the highest quality raw materials.

  • Autoclavable
  • Available in a variety of sizes/colors
  • Manufactured to yield exceptional dimensional stability
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Outer 20 mm
Material Rubber
Product Type Straight Plug Stopper
For Use With (Equipment) Vacule Ampules
Color Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.