Learn More
DWK Life Sciences Wheaton™ Unlined Aluminum Center Disc Tear-Out Seals
GREENER_CHOICESDP
Catalog No. 0645176 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Red
Closure Size:
13 mm
20 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Size
06-451-76 Blue 20 mm
06-451-73 Blue 13 mm
06-451-74 Red 13 mm
06-451-77 Red 20 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-451-76 Supplier DWK Life Sciences Supplier No. 22418305
Add to Cart
Add to Cart
Exposes the top of the rubber stopper to the syringe when the center disc is opened or removed
  • Available in 13 or 20mm O.D.
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Aluminum
Quantity 1000/Cs.
Closure Size 20 mm
Product Type Center Disc Tear-out Cap