Learn More
DWK Life Sciences Wheaton™ Unlined Aluminum Open-Top Seals
SDP
Catalog No. 1112617 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Closure Size:
11 mm
13 mm
20 mm
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Size
11-126-17 Natural 11 mm
11-126-25 Blue 20 mm
11-126-26 Red 20 mm
11-126-22 Blue 13 mm
11-126-23 Red 13 mm
11-126-18 Blue 11 mm
11-126-27 Green 20 mm
06-449-21 Natural 13 mm
11-126-19 Red 11 mm
11-126-20 Green 11 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 11-126-17 Supplier DWK Life Sciences Supplier No. 22417601
Add to Cart
Add to Cart
Open-top seal exposes stopper for sample retrieval with a syringe
  • Unlined
  • Closure Type: Seal
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material Aluminum
Quantity 1000/Cs.
Closure Size 11 mm
Product Type Open Top Cap