Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps with Foam/LDPE Liners
Shop All DWK Life Sciences Products

Description
- Lined with foam polyethylene
- Excellent barrier to aqueous solutions
- Repeat sealability
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| No. per Case | 200 Cs. |
| Liner Material | Foam/LDPE |
| Neck Finish | 28-400 |
| Certifications/Compliance | ISO 9001 |
| For Use With (Equipment) | Glass bottle or vial |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.