Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps with Polyvinyl Liners
Shop All DWK Life Sciences Products

Description
- White polypropylene screw cap provides clear surface for writing on cap
- Poly-vinyl liner provides an excellent moisture barrier
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| No. per Case | 72 Cs. |
| Neck Finish | 43-400 |
| Certifications/Compliance | ISO 9001 |
| Product Type | Polypropylene Cap |
| For Use With (Equipment) | Round bottle |
| Closure Liner | Polyvinyl |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.