Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yosemite Conservancy Where's Rodney by Carmen Bogan and Floyd Cooper
SCI_ED

Catalog No. S000943 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Subject:
Nature, Parks
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Subject
S000943 Nature, Parks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000943 Supplier Yosemite Conservancy Supplier No. 9781951179052
Add to Cart
Add to Cart

Literature for educational purposes

  • Educational Book

Specifications

Grade 1
Format Paperback Book
Product Type Paperback, Parks
Subject Nature, Parks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.