Learn More
International Enviroguard™ White Biohazard Coveralls with Attached Boots
Shop All International Enviroguard Systems ProductsDescription
- Durable fabric is built to withstand tears and abrasions in tough working environments
- Both fabric and seams pass ASTM F1670 and F1671 tests for protection against blood, bodily fluids, and bloodborne pathogens
- Zipper front with a sealable storm flap helps keep contaminants from entering or seeping through otherwise-exposed zipper seams
- Reinforced with double-sided tape at the wrists, your gloves will remain secure as you work
- Red fabric pull tab on the zipper makes it quick and easy to find so you can suit up or down in no time
- Taped seams seal off seams from contaminants, while elastic wrists and ankles keep coveralls secure and in place to avoid skin exposure
- For added convenience, thumb and finger loops are included to help prevent sleeves from riding up
- Attached boots protect your feet while latex-free, elastic wrists offer a secure fit to prevent skin exposure of the arms
- Mandarin collar with hook and loop closure at neck provides a better neck seal
- Roomy, one-piece raglan sleeves resist tears around shoulders
- Gusseted crotch gives the user more freedom to move, bend, crouch, climb, and squat without ripping fabric
- Possible applications include emergency response, healthcare, category 3 black water, sanitation work, crime scene remediation, and animal research
Specifications
Specifications
| Product Type | A50 Breathable Particle and Splash Protection Coveralls |
| Size | Medium |
| Gender | Unisex |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic at Back |
| Ankle Style | Attached Boots |
| Cuff Style | Elastic, Finger Loops |
| Neck Style | Collared |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.