Learn More
International Enviroguard™ White Biohazard Suit with Elastic Wrists and Ankles
Shop All International Enviroguard Systems ProductsDescription
- Both the fabric and seams pass ASTM F1670 and F1671 tests for protection against blood, bloodborne pathogens, and bodily fluids
- Durable fabric is built to withstand tears and abrasions in tough working environments
- Zipper front with a sealable storm flap helps keep contaminants from entering or seeping through exposed zipper seams
- Red fabric pull tab on the zipper makes it quick and easy to find so you can suit up or down in no time
- Taped seams help keep contaminants out for added protection, while elastic wrists and ankles keep coveralls secure and in place
- Convenient thumb and finger loops help keep sleeves from riding up while you work
- Double-sided tape around wrists helps keeps gloves in place during donning
- Storm flap over zipper helps prevent contaminants from entering through otherwise exposed zipper seams
- Roomy one-piece raglan sleeves resist tears around shoulders
- A gusseted crotch gives the user more freedom to squat, bend, and crouch without ripping fabric
- Possible applications include emergency response, healthcare, category 3 black water, crime scene remediation, and animal research
Specifications
Specifications
| Product Type | A50 Breathable Particle and Splash Protection Coveralls |
| Size | X-Large |
| Gender | Unisex |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic at Back |
| Ankle Style | Elastic |
| Cuff Style | Elastic, Thumb Loop |
| Neck Style | Collared |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.