Learn More
Description
- Men's Performance Work Socks provide support and protection for your feet while you work.
- The mesh upper foot is designed for ventilation and moisture evaporation, keeping your feet dry throughout the day.
- Socks feature reinforced heel and toe as well as an Achilles heel pad for added protection and comfort.
- The socks non-binding top do not slip down and provide all-day gentle comfort.
- Added compression at the arch provides added support and stability.
Specifications
Specifications
| Size | X-Large |
| For Use With (Application) | Cleanroom Use |
| Material | Polyester, Nylon, Spandex |
| Color | White, Blue |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.