Learn More
Leica Biosystems WHITE CASSETTES CS/1000WHITE
Supplier: Leica Biosystems 38440200
IP C Cassettes III are one-piece processing cassettes with a 35 degree writing surface. The cassettes are taped for easy loading into ink printers. Available in eleven colors in quantities of 1000.