Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ White Dry Field Mapping Paper
SCI_ED

Catalog No. S27750 Shop All Eisco Products
Change view
Click to view available options
Class Size:
100 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S27750 100 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27750 Supplier Eisco™ Supplier No. PH0918DFMWHT
Add to Cart
Add to Cart

White Dry Field Mapping Paper is non-conductive paper for plotting electric fields and electrode configurations.

  • Contains 20 x 25 cm grid for plotting electric fields (20 Grids/Page)
  • For use with other field mapping equipment
  • Bound pad includes 100 sheets
  • Sheet Dimensions (L x W): 11 x 8.5 in.

Specifications

Grade 7 to Undergraduate
Height (English) 11 in.
For Use With (Application) Plotting Electric Fields and Electrode Configurations
Includes 100 Sheets of Non-conductive Paper
Quantity 100 Pk.
Height (Metric) 27.9 cm
Class Size 100 Students
Product Type Non-conductive Paper
Description Dry Field Mapping Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.