Description
These jars provide an evaporation and contamination-free mixing vessel to enable a manufacturing process in an oxygen-reduced space. After mixing the jars can be used to dispense into specific dispensing containers or directly to the patient.
- Type: Stirrer Part
Specifications
Specifications
| Capacity (Metric) | 30 mL |
| Color | White |
| Quantity | 10/Pk. |
| Type | Stirrer Part |