Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ White Metal Plastisol Lined Caps

Catalog No. 02993406
Change view
Click to view available options
Neck Finish:
38-400
48-400
53-400
58-400
63-400
70-400
89-400
No. per Case:
144 Cs. (12/Pk.)
1900 Cs.
2300 Cs.
288 Cs. (12/Pk.)
3800 Cs.
576 Cs. (12/Pk.)
576 Cs. (144/Pk.)
700 Cs.
Packaging:
Bagged
Bulk
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case Packaging
02-993-406 38-400 576 Cs. (144/Pk.) Bagged
02-993-398 53-400 288 Cs. (12/Pk.) Bagged
02-991-106 58-400 1900 Cs. Bulk
02-993-372 70-400 144 Cs. (12/Pk.) Bagged
02-993-340 58-400 576 Cs. (12/Pk.) Bagged
02-991-128 38-400 3800 Cs. Bulk
02-993-404 89-400 144 Cs. (12/Pk.) Bagged
02-993-415 58-400 288 Cs. (12/Pk.) Bagged
02-993-308 63-400 288 Cs. (12/Pk.) Bagged
02-991-172 48-400 2300 Cs. Bulk
02-991-092 89-400 700 Cs. Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 02-993-406 Supplier Qorpak™ Supplier No. CAP00442

Tolerates wide ranges of temperature

  • Sturdy and unbreakable
  • Impact resistant
  • Made of steel coated with anti-corrosive tinplate
  • Permit hot fill operations to produce vacuum seal
  • Provide resistance to mild acids as in food products

Specifications

Color White
Material Metal
No. per Case 576 Cs. (144/Pk.)
No. per Pack 576
Packaging Bagged
Sterility Non-sterile
Liner Material Plastisol
Neck Finish 38-400
Disposable Reusable
Product Type Threaded Screw Cap
For Use With (Equipment) Wide mouth bottle
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.