Learn More
Valutek™ White Polyethylene Film Adhesive Roller
Catalog No. 191502130
Change view
Click to view available options
No. per Case:
4 Rolls/Cs.
Catalog No. No. per Case
19-150-2130 4 Rolls/Cs.
1 options
Catalog No. 19-150-2130 Supplier Valutek™ Supplier No. VTTRLRW18
Add to Cart
Add to Cart
Manufactured with polyethylene and acrylic solvent based adhesive sheets. The perforated sheets allow for removal after use, providing a new surface area for the next application.
  • A bactericide is used to prevent microbial growth
  • This product is recommended for use in a cleanroom Class 100-1,000(ISO 5-6) critical environment that requires high contamination control and cleanliness
  • It is designed to effectively capture dirt and dust from smooth surfaces such as floors, walls, and ceilings in controlled environments
  • All Valutek™ adhesive rollers are tested and manufactured in ISO compliant facilities under Valutek™ inspection and stringent process control to ensure Valutek™ quality standard and product specification compliance
Specifications
For Use With (Application) Smooth surfaces such as floors, walls, and ceilings in controlled environments (to capture dirt and dust)
Includes 107 18 in. Sheets/Roll
No. per Case 4 Rolls/Cs.
Product Type Polyethylene Film Adhesive Roller