Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ Caps for Screwthread Diagnostic Vials
SDP

Catalog No. 12141620 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
15.5 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
12-141-620 15.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-141-620 Supplier Wheaton™ Supplier No. 300180
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For the safe and secure closure of vials in the laboratory. Wheaton™ Caps for Screwthread Diagnostic Vials are manufactured from pharmaceutical grade polypropylene.

  • White in color
  • To fit thread finish 15.5mm (15.5R4)
TRUSTED_SUSTAINABILITY

Specifications

Closure Size 15.5 mm
Description Cap
Material Polypropylene
Color White
Quantity 1000 Pack
For Use With (Equipment) Screw Thread Diagnostic Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.