Learn More
United Scientific White Streak Plates, 1 x 2 in.
SDPSCI_ED
Catalog No. S81055 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
10/Pk.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S81055 12/Pk.
S81053 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S81055 Supplier United Scientific Supplier No. PSP001W
Add to Cart
Add to Cart
Streak plates designed for hardness testing of rocks and minerals.
  • Excellent for Hardness Testing of Rocks and Minerals
  • White porcelain streak plate
  • Size: 1 x 2 in.
Specifications
Quantity 12/Pk.
For Use With (Application) Rock and Mineral Identification
Product Type Streak Plate
Dimensions (L x W) 1 x 2 in.