Learn More
Description
- Tyvek™ tags are a lightweight, tear proof and weather resistant alternative to cardstock tags
- They provide more durability but still offer the flexibility to write your own custom message on the spot
- Metal grommet prevents tearing from the point of attachment
- Tag includes a write-on area that accepts most marking pens
- Tags are appropriate for indoor or outdoor use
Specifications
Specifications
| Quantity | 1000/Pk. |
| Color | White |
| Material | Tyvek |
| Shape | Rectangle |
| Dimensions (L x W) | 2.875 x 5.75 in. |
| Label Type | Not laminated |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.