Learn More
Description
- For disposal of capillary tubes, pipette tips, stir rods and other point-of-use items
- Maximize workspace with hanging biohazard disposal units
- Made of sturdy plastic-lined paperboard
- Solid-bottom design improves rigidity and waste capacity
- Safe-Keeper can be autoclaved or incinerated
- Accommodates up to 1100cc of material and comes with a Hook-Up™ self-adhesive hanger
- Made in USA
Specifications
Specifications
| Capacity (English) | 1100 cc |
| Dimensions (L x W x H) | 5 x 2 x 11 in. (12.7 x 5.1 x 27.9 cm) |
| Disposable | Single-use |
| Height (Metric) | 27.9 cm |
| Length (Metric) | 12.7 cm |
| Width (Metric) | 5.1 cm |
| Autoclavable | Autoclavable |
| Liner Material | Plastic |
| For Use With (Application) | Disposal of capillary tubes, pipette tips, stir rods and other point-of-use items |
| Width (English) | 2 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.