Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Whizard™ Cut-Resistant Heavy-Duty Slipguard Gloves, Left Hand

Catalog No. 1140026C Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11-400-26C Small
11-400-26E Medium
11-400-26G Large
11-400-26J X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11-400-26C Supplier Wells Lamont™ Supplier No. 133554
This item is not returnable. View return policy

Whizard™ Cut-Resistant Heavy-Duty Slipguard Gloves are standard pattern gloves that use a unique combination of ultra-high strength fibers and stainless steel to provide hand protection against cuts and slashes. Polyurethane grip pattern provides a no-slip grip.

  • Fabric consists of high-performance fibers and stainless-steel core to provide an excellent source of cut resistance
  • Polyurethane pattern covering palm, fingers and thumb crotch improves gripping
  • Ideal for the pulp and paper industry as well as for plastic trimming
  • Also features excellent thermal properties
  • 3 in. (7.6 cm) ribbed cuffs
  • Overedge color indicates size: red (XXS), green (XS), yellow (S), blue (M), white (L), orange (XL)
  • Made in US

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance ASTM F2992-15
Coating Material Polyurethane
Color Green
Cut Resistance Rating ANSI A5
Hand Left
Latex Free Yes
Material Fibers, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Pulp and paper industry and plastic trimming
Glove Cuff Color Green
Glove Cuff Style Continuous Knit
Hem Color Yellow
Model 133554
Series Whizard™ Slipguard™ Standard Pattern
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.