Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Whizard™ Defender™ 10 Cut-Resistant Gloves

Catalog No. 19167806A Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-167-806A X-Small
19-167-806D Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-167-806A Supplier Wells Lamont™ Supplier No. 135478
Add to Cart
Add to Cart
This item is not returnable. View return policy

Whizard™ Defender™ 10 Cut-Resistant Gloves with high performance fibers and stainless steel combination offer ANSI Cut Level A6 cut resistance. 10-gauge gloves are ideal for handling sharp surfaces, food preparation, and meat and poultry processing.

Patented combination of high-performance fibers and stainless steel creates superior cut resistant properties, eliminating the high cost of lacerations.

  • Standard cuff
  • Ambidextrous
  • Prewashed and preshrunk
  • 10 gauge: ANSI A6
  • Made in the USA

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance FDA CFR 21, ASTM F2992-15, ASTM D3884-09
Coating Material Uncoated
Color White, Blue
Cut Resistance Rating ANSI A5
Hand Ambidextrous
Latex Free Yes
Material Fibers, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size X-Small
For Use With (Application) Medium duty applications
Glove Cuff Color Blue
Glove Cuff Style Plain
Hem Color Green
Model 135478
Series Whizard™ Defender™ 10
Yarn Gauge 10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.