Learn More
Wells Lamont™ Whizard™ Metal Mesh Hand Gloves
Catalog No. 17200385 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-385 Small
17-200-384 X-Small
17-200-386 Medium
17-200-387 Large
17-200-388 X-Large
17-200-389 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-200-385 Supplier Wells Lamont™ Supplier No. CM030002
Add to Cart
Add to Cart
This item is not returnable. View return policy
Whizard™ Metal Mesh Hand Gloves protect against cuts and feature micro plasma welded rings with a self-tensioning wrist closure.
  • Metal rings are micro plasma welded to prohibit the growth of bacteria
  • Patented self-adjusting wrist tensioning system with no straps or buckles to fasten
  • Metal serial number tag
  • Ambidextrous
  • Easy to clean
Specifications
Hand Ambidextrous
Latex Free Yes
Material Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Food, Beef, Chicken, and Pork Processing, Restaurant and Steel Industry
Glove Cuff Color White
Model CM030002
Series Whizard™ Metal Mesh