Description
- Metal rings are micro plasma welded to prohibit the growth of bacteria
- Patented self-adjusting wrist tensioning system with no straps or buckles to fasten
- Metal serial number tag
- Ambidextrous
- Easy to clean
Specifications
Specifications
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Stainless Steel Fiber |
| Product Type | Cut-resistant Gloves |
| Size | X-Large |
| For Use With (Application) | Food, Beef, Chicken, and Pork Processing, Restaurant and Steel Industry |
| Glove Cuff Color | Yellow |
| Model | CM030005 |
| Series | Whizard™ Metal Mesh |