Learn More
Wells Lamont Industrial™ Whizard™ Z40 Armguard
Catalog No. 18063970 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Cuff Style:
Tube Top
Wide Top
Length (English):
18 in.
22 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cuff Style Length (English)
18-063-970 Tube Top 22 in.
18-063-969 Wide Top 22 in.
18-063-971 Tube Top 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-063-970 Supplier Wells Lamont Industrial™ Supplier No. 135658
Add to Cart
Add to Cart
Whizard™ Z40 cut resistant sleeve. Offers level ANSI A6 cut resistance. Made in the USA.
Whizard™ Z40 armguard offers superior cut protection from arm lacerations. This armguard is an ideal complement to Whizard™ hand protection.
  • Constructed from a combination of high performance materials, Z40 armguards offer ANSI level A6 cut protection.
  • The unique knit structure allows greater circulation of air, keeping arms cool and comfortable while working.
  • The armguard offers the same material composition in both a standard and tube version.
  • The standard version offers a wide upper arm opening for bigger biceps.
  • Comes with a clip to keep the sleeve in place on the upper arm.
Specifications
Size Standard
Product Type Cut-resistant Sleeve
Material HPPE
Color Blue
Sterility Non-sterile
Style Elastic at Both Ends
Disposable Reusable
Closure Type Elastic Ends
Seam Type Seamless
Cuff Style Tube Top
Latex Free Yes
Length (English) 22 in.