Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies UniStore™ Amber Reagent Bottles, Wide Mouth, HDPE
SDPSCI_ED

Catalog No. S82949 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S82949 30 mL
S82950 60 mL
S82951 125 mL
S82952 250 mL
S82953 500 mL
S82954 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. S82949 Supplier United Scientific Supplies Supplier No. 33461
Add to Cart
Add to Cart
This item is not returnable. View return policy

Wide Mouth Amber Reagent Bottles are made from HDPE and protect samples with light sensitivity.

  • United Scientific™ Wide Mouth, HDPE, reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant.
  • The amber bottles are designed to reduce UV light transmission to protect light-sensitive contents and conform to USP requirements for storage of light sensitive materials.
  • Excellent chemical resistance to most acids, bases, and alcohols.
  • Polypropylene caps are included.
  • Protect light-sensitive materials during shipping and storage.
  • Meet USP requirements for storage of light-sensitive materials.
  • Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact.
  • Free of heavy metals, with Leakproof closure with integrated seal ring.
  • Precision molded for consistent wall thickness and smooth interior and exterior surfaces.

Specifications

Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Amber
Material HDPE
Quantity 72 Pk.
Sterility Non-sterile
Product Line UniStore
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide
Shape Round
Height (Metric) 63 mm
Product Type Amber Bottle
Height (English) 2.5 in.
Closure Material Polypropylene
Closure Size 28-415
Through the Closure Dispensing No
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.