Learn More
United Scientific Supplies UniStore™ Amber Reagent Bottles, Wide Mouth, HDPE

Shop All United Scientific Supplies, Inc Products


Description
- United Scientific™ Wide Mouth, HDPE, reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant.
- The amber bottles are designed to reduce UV light transmission to protect light-sensitive contents and conform to USP requirements for storage of light sensitive materials.
- Excellent chemical resistance to most acids, bases, and alcohols.
- Polypropylene caps are included.
- Protect light-sensitive materials during shipping and storage.
- Meet USP requirements for storage of light-sensitive materials.
- Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact.
- Free of heavy metals, with Leakproof closure with integrated seal ring.
- Precision molded for consistent wall thickness and smooth interior and exterior surfaces.
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Amber |
| Material | HDPE |
| Quantity | 12 Pk. |
| Sterility | Non-sterile |
| Product Line | UniStore |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Height (Metric) | 168 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.