Learn More
Anaerobe Systems ATTMSP - Anaerobic Tissue Transport Medium Surgery Pack
SDP
Supplier:  Anaerobe Systems AS915
Encompass
This item has a minimum qty of 2 per supplier requirements.
Anaerobic Tissue Transport Medium Surgery Pack (ATTMSP) is a mineral salt based semi-solid media in a wide mouth tube. Designed as a non-nutrituve holding medium, it maintains viability of microorganisms through collection & transport of clinical specimens. This media is prepared, dispensed, & packaged under oxygen-free conditions. Each tube is individually packaged in sterile autoclaveable surgery packs.ATTMSP contains buffered mineral salts in a semi-solid media, w/ sodium thioglycollate & cysteine added as reducing agents. This medium maintains the viability of most microorganisms without multiplication & allows for dilution of inhibitors present in clinical material, & meets the viability requirements of obligate anaerobes. Tubes are 25mm wide to facilitate placement & removal of larger sized samples such as tissue biopsies or feces. Resazurin is added as an indicator of significant oxygen exposure to the media.
Catalog No. NC9647836
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.