Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Wide Mouth Plastic Reagent Bottle
SCI_ED

Catalog No. S24490 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S24490 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24490 Supplier Eisco™ Supplier No. CH0172EW
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Wide mouth reagent bottles made of rigid, chemically-resistant polypropylene with a wide mouth.

  • Industrial quality reagent bottle with a 1000 mL capacity
  • Rigid polypropylene construction provides excellent chemical resistance to acids, alcohols and alkalis, and very good resistance to aldehydes and hydrocarbons
  • Stable at temperatures between -20°C and 135°C
  • Includes a leak-proof screw cap
  • Both bottle and cap are translucent with excellent optical clarity
  • Bottle features a wide mouth
  • Outer diameter of mouth measures 52 mm and inner diameter measures 44 mm
  • Overall size: 8.7 in. tall and 3.4 in. diameter
  • Bottle and screw cap are autoclavable and top rack dishwasher safe for easy cleaning and sterilization

Specifications

Capacity (Metric) 1000 mL
Color Translucent
Material Polypropylene
Length (Metric) 9.52 cm
Quantity 1
Sterility Non-sterile
Width (Metric) 9.52 cm
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Height (Metric) 22 cm
Height (English) 8.7 in.
Through the Closure Dispensing No
Autoclavable Autoclavable
Capacity (English) 33.81 oz.
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.