Learn More
Eisco™ Wide Range pH Test Strips 11.5-14, 3-Pad, Quick Read, Premium Quality, 50 Single Use Strips in Bag
SCI_ED
Catalog No. S02934 Shop All Eisco Products
Change view
Click to view available options
pH Range:
11.5 to 14
Catalog No. pH Range
S02934 11.5 to 14
1 options
Catalog No. S02934 Supplier Eisco™ Supplier No. CH201804
Add to Cart
Add to Cart
Eisco Labs wide range pH test strips 11.5 to 14 feature a 3-pad design for quick and accurate readings. Premium quality, with 50 single-use strips included in a convenient bag. Ideal for diverse pH testing needs.
  • Quick and precise pH measurement: Each strip measures pH levels from 11.5-14 in increments of 1.0 unit, providing rapid and accurate testing
  • Versatile application: Suitable for use in various fields such as chemistry, biology, agriculture, water quality, and environmental sciences
  • Triple pad accuracy: Features three pads per strip, offering a wider detection range and reliable results in just seconds
  • User friendly design: Packaged with a vivid color chart for easy pH determination
  • Traceability: Each bottle is marked with a lot number and expiry date for easy reference
  • Discover ease and efficiency with our Wide Range pH Test Strips, tailored for quick and accurate pH assessments across diverse materials
  • Ideal for educational, industrial, and environmental monitoring applications, these strips ensure you get dependable results fast
  • Included in this package are 100 pH test strips, each capable of discerning pH levels from 11.5 to 14 with clear color changes
  • Housed in a handy flip-up vial, the strips are designed for easy access and straightforward reading with the help of the color chart provided on the bottle
  • Please note, these strips are not intended for medical use and should not be used for diagnosing or treating health conditions
Specifications
pH Range 11.5 to 14
Type Wide Range pH Test Strips 11.5-14