Learn More
Thermco Products Wifi Vaccine Logger A
Catalog No. 04500351
Change view
Click to view available options
Display Type:
LCD
Catalog No. Display Type
04-500-351 LCD
1 options
Catalog No. 04-500-351 Supplier Thermco Products Supplier No. UTREL30WIFISP2
Add to Cart
Add to Cart
This CDC and VFC compliant, ISO/IEC 17025 NIST-traceable 30 day temperature data logger with a display for monitoring vaccine storage in refrigerators or freezers.
  • This data logger meets or exceeds CDC guidelines.
  • Large, easy-to-read display shows current, minimum, and maximum temperatures along with alarm status for up to 30 days at a glance.
  • Non-volatile memory stores up to 16,129 temperature readings.
  • Statistical memory tracks maximum and minimum temperatures and alarm durations for each of the last 30 days, ensuring robust monitoring.
  • Automatically uploads real-time temperature data to LogTag Online via WiFi.
  • PDF data download option is available without software when WiFi is unavailable.
  • External probe measures temperatures from -40°C to 99°C (-40°F to 210°F), supporting accurate vaccine temperature monitoring.
Specifications
Type Temperature Datalogger
Display Type LCD
Alarm Types Audible, LED Indicator