Learn More
Thermco Products Wifi Vaccine Temperature Logger
Catalog No. 04500350
Change view
Click to view available options
Model:
Temperature Logger A
Temperature Logger B
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
04-500-350 -
04-500-341 Temperature Logger A
04-500-342 Temperature Logger B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 04-500-350 Supplier Thermco Products Supplier No. UTRED30WIFIWSP2
Add to Cart
Add to Cart
This CDC and VFC compliant, ISO/IEC 17025 NIST-traceable 30 day temperature data logger with a display for monitoring vaccine storage in refrigerators or freezers.
  • This CDC and VFC compliant, ISO/IEC 17025 NIST-traceable 30 day temperature data logger with a display for monitoring vaccine storage in refrigerators or freezers.
  • Automatically uploads real-time temperature data to a LogTag Online cloud account via WiFi.
  • Non-volatile memory stores up to 16,129 temperature readings.
  • Statistical memory tracks maximum and minimum temperatures and alarm durations for each of the last 30 days.
  • Users can view and reset minimum and maximum readings at any time.
  • Audible and flashing LED alerts provide immediate notification of temperature excursions.
  • Data can be downloaded using LogTag Analyzer software or generated as a PDF without software if WiFi is unavailable.
  • External buffer bottle probe measures temperatures from -40°C to +99°C (-40°F to +210°F) for accurate vaccine monitoring.
Specifications
Type Temperature Datalogger
Display Type LCD
Alarm Types Audible, Visual, SMS, Voice