Learn More
Thermo Scientific Chemicals Wig-L-Bug™ Accessories
Catalog No. AA44531KT
Change view
Click to view available options
Quantity:
100/Pk.
5/Pk.
Each
Product Type:
Vial
Vial Adapter
Height (English):
2 in.
1 ft.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Height (English)
AA44531KT Each Vial Adapter -
AA44530KN 100/Pk. Vial 2 in.
AA42628KJ 5/Pk. Vial 1 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA44531KT Supplier Thermo Scientific Chemicals Supplier No. 44531KT
For use with Digital Wig-L-Bug Mixing/Grinding Mill
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Polystyrene vial with polyethylene slip-on cap; 1 ft. long x 0.5 in. dia.
  • Polystyrene vial with polyethylene slip-on cap; 2 ft. long x 0.5 in. dia.
  • Adapter for 2 in. long, 0.5 in. diameter vials
TRUSTED_SUSTAINABILITY
Specifications
Description Accessory for Digital Wig-L-Bug mixing/grinding mill
Quantity Each
Product Type Vial Adapter