Learn More
Selleck Chemical LLC WIKI4
SDP
Supplier:  Selleck Chemical LLC S74901G
Encompass_Preferred
WIKI4 is a novel Tankyrase inhibitor with IC50 of 15 nM for TNKS2 and leads to inhibition of Wnt/beta-catenin signaling
Catalog No. 50-000-01423
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.