Learn More
Eisco™ Wilberforce Pendulum
SCI_ED
Catalog No. S23773 Shop All Eisco Products
Change view
Click to view available options
Material:
Steel
Catalog No. Material
S23773 Steel
1 options
Catalog No. S23773 Supplier Eisco™ Supplier No. WILBF
Add to Cart
Add to Cart
Includes
  • (1) Adjustable Wilberforce Pendulum Mass
  • (1) Helical Spring
  • (1) Mounting Rod connected to boss head clamp
For demonstration of coupled mechanical oscillation.
  • Pendulum made from steel
  • Radial arms with threaded adjustable weight for moment of inertia to be tuned to torsional vibration period
  • Device needs to be mounted 1m from surface
  • For executive desk or advanced physics demonstration
Specifications
Product Type Pendulum, Wilberforce
Material Steel
For Use With (Application) Demonstration of coupled mechanical oscillation
Includes Adjustable Wilberforce pendulum mass, helical spring, mounting rod connected to boss head clamp