Learn More
Leica Biosystems Leica Wild M600 Series replacement bulb - 12 V 50 W halogen (MPN 10384643)
Supplier: Leica Biosystems 10384643
Replacement halogen bulb for Leica Wild M600-series microscopes. This 12 V, 50 W T-3.5 lamp provides a compact, high-intensity light source for surgical and stereo microscope illumination assemblies. OEM part number 10384643; compatible with M651, M600, M680, and M690 models.
- Provides bright, high-intensity illumination suitable for microscope lighting.
- 12 V, 50 W halogen output for consistent lamp performance.
- T-3.5 form factor in a compact bulb design.
- Designed for installation in standard microscope lamp housings.
- Direct replacement intended to restore original light output.