Learn More
Ansell WildCat™ Heat-Resistant Goggles
Catalog No. 19049922 Shop All Ansell Products
Change view
Click to view available options
Lens Tint:
Amber
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-049-922 Amber
19-049-921 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-049-922 Supplier Ansell Supplier No. 20527
This item is not returnable. View return policy
Offer extreme heat resistance
  • Will not melt, drip, or ignite at 350° for five minutes
  • Adjustable side vent to ensure increased airflow
  • Fits naturally to face contours to block out airborne contaminants
  • Lightweight and comfortable with a pivot headband that allows optimal placement
  • Antifog lenses
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, UV-treated
Lens Type Splash protection
Certifications/Compliance ANSI Z87.1-2010
Lens Tint Amber
Product Type Heat-resistant safety goggles
For Use With (Application) Splash, heat and dust protection
Ventilation Details Adjustable side vent
Lens Material Polycarbonate
UV Protection 0.999