Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Williams and Briggs Salt Mixture

Catalog No. MP290283710 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 kg
10 kg
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP290283710 10 kg
MP290283702 2 kg
MP290283750 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP290283710 Supplier MP Biomedicals™ Supplier No. 0290283710
Add to Cart
Add to Cart

Salt mixture for research animal diet.

  • Salt Mixture
  • For research animal diet
  • Powder
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
For Use With (Application) Research animal diet
Quantity 10 kg
Storage Requirements Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.