Learn More
Sigma Aldrich Fine Chemicals Biosciences Williams Medium, Cell Culture Tested | N/A | 500 ML Bottle
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W0397500ML
Encompass
Williams Medium E cell culture medium | Contains essential: nutrients, vitamins, amino acids, inorganic salts | Buffered with: sodium bicarbonate | Without phenol red pH indicator | Sterile-filtered | Requires serum or growth factor supplementation | For use in 5% CO2 cell culture incubators | Supports growth of various mammalian cell lines | Volume: 500mL | CAS No: N/A | Unit of Measure: 500mL bottle
Catalog No. NC2602148
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.