Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ Disposable NMR Tube Caps, 4.0mm Dia., Blue

Catalog No. 16800215 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Includes:
100 per pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
16-800-215 100 per pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-800-215 Supplier Bel-Art™ Supplier No. 660040101

Different colors help track samples based on various reference solvents.

Alert: When using Chloroform or Acetone as a reference solution, please avoid direct contact between the sample and the disposable cap, and avoid long term storage over 2 hours. If these conditions cannot be avoided, please use PTFE tube caps.

Specifications

Includes 100 per pack
Diameter (Metric) Outer 4.0 mm
Description Model: Wilmad™ 521-G-100
Material Polyethylene
Color Blue
For Use With (Application) NMR Spectroscopy
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.