Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Doty™ Susceptibility Plugs

Catalog No. 16800149 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Material:
Aurum™
PPS
Length (Metric):
8 mm
9 mm
14 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Length (Metric)
16-800-149 PPS 8 mm
16-800-145 Aurum™ 14 mm
16-800-146 Aurum™ 9 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16-800-149 Supplier Bel-Art™ SP Scienceware™ Supplier No. 660000049

missing translation for 'validMainframeMsg'

Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Doty™ Susceptibility Plugs allow the sample to be substantially concentrated.

  • Match a wider range of solvents
  • Less fragile in variable temperature experiments involving freeze-pump-thaw
  • Use with our 3, 5 or 8 mm thin wall tubes
  • Easy sample loading and cleaning
  • Fully compatible with Bruke™ Agilent™ (formerly Varian™) and JEOL™ spectrometer/probe
  • Designed for easier air bubble removal.
  • May be removed from the sample tube with minimal sample loss (1-2%) after spectra are obtained, then used with other samples

Specifications

Product Type Susceptibility Plugs
Material PPS
Product Line Model: SP-PS-3
Length (Metric) 8 mm
For Use With (Equipment) 5 mm Wilmad-LabGlass Thin Wall Precision Tubes

Store plugs in deuterated solvent to suppress water absorption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.