Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Spinner Bearing
Catalog No. 16800142 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Model:
SB-5-7
SB-8-7
Diameter (Metric) Outer:
For tubes 5mm O.D.
For tubes 8mm O.D.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Diameter (Metric) Outer
16-800-142 SB-5-7 For tubes 5mm O.D.
16-800-143 SB-8-7 For tubes 8mm O.D.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-800-142 Supplier Bel-Art™ Supplier No. 660000042
Ensure shimming quality of your glass tubes. Bel-Art SP Scienceware™ Wilmad™ Spinner Bearing will determine which of your NMR tubes are warped and bent.
  • Useful in labs that wash, dry and reuse precision NMR tubes
  • Warped tubes will bind in the spinner bearing while good tubes spin freely
  • Keep a tube tester beside every NMR spectrometer for easy access
Specifications
For Use With (Equipment) Quartz, Supracil, or Precision tubes
Diameter (Metric) Outer For tubes 5mm O.D.
Model SB-5-7