Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Wimshurst Generator
SDPSCI_ED

Catalog No. S27877 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
For Use With (Application):
Demonstrating High Voltage Electric Charges
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S27877 Demonstrating High Voltage Electric Charges
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27877 Supplier United Scientific Supplies Supplier No. WIMS02
Add to Cart
Add to Cart

For demonstrating high voltage electric charges

  • Classroom-friendly
  • Generates large sparks for high voltage demonstrations
  • Features acrylic rotating disks, quality foil strips/brushes, insulated cylindrical capacitors, and a durable plastic platform
  • Activity guide included

Specifications

Product Type Wimshurst Generator
For Use With (Application) Demonstrating High Voltage Electric Charges
Includes Acrylic Rotating Disks, Quality Foil Strips/Brushes, Insulated Cylindrical Capacitors, Durable Plastic Platform, and an Activity Guide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.