Learn More
Cayman Chemical (+)-WIN 55,212-2 (mesylate), 131543-23-2, 5 mg
SDP
Supplier:  Cayman Chemical 100090235
Encompass_Preferred
A potent aminoalkylindole CB receptor agonist (Ki = 62.3 and 3.3 nM for human recombinant CB1 and CB2 receptors, respectively); increases extracellular glutamate levels in primary cultures of rat cerebral cortex neurons, displaying a bell-shaped concentration-response curve; induces release of the proinflammatory neuropeptide CGRP from trigeminal ganglion neurons in a calcium-dependent manner (EC50 = 26 µM)
Catalog No. NC0403209
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.