Description
- New version of an old favorite has 12 colorful plastic vanes and a balanced tail
- Connect to DC generator and light bulb to indicate electrical output with the included leads
- Clamps to lab bench
Specifications
Specifications
| For Use With (Application) | Study aerodynamic transfers by generating waves by connecting to DC generator and light bulb to indicate electrical output with the included Leads |
| Includes | 12 Colorful Plastic vanes and a balanced tail |