Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Wiper

Catalog No. 19104454 Shop All Brady Corporation Products
Catalog No. 19-104-454 Supplier Brady™ Supplier No. BPACLFILM2

Use printhead cleaning Wipers to keep your thermal transfer printhead free of dust and dirt.

  • Printhead cleaning film provides an easy solution to a common problem by keeping your printer in top condition
  • Blue grit sheets are designed to effectively remove dirt and other contaminants from the print head within a thermal transfer printer
  • Printhead cleaning film measures 2 in. W x 6 in. H
  • Cleans printheads safely and easily

Specifications

Product Type Cleaning Kit - Wiper
For Use With (Equipment) Brady Label Printers
Width (Metric) 50.8 mm
Dimensions (L x W) 15.24 x 5.08 cm (W x H)
Sterility Non-sterile
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.