Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Wire Epoxy-Coated Test Tube Rack
SDPSCI_ED

Catalog No. S43723 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Holds:
Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
S43723 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43723 Supplier United Scientific™ Supplier No. TTWE01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heavy Wire Test Tube Rack features a durable epoxy coating, and can hold up to 36 tubes.

  • Heavy wire test tube rack.
  • Durable epoxy coating.
  • Holds up to 36 tubes.

Specifications

Autoclavable Autoclavable
Color Blue
Holds Tubes
Material Wire
Positions 36
Shape Cubical
Stackable Yes, When Empty
Sterility Non-sterile
For Use With (Equipment) Test Tubes or Culture Tubes
Length (Metric) 156 mm
Includes Test Tube Rack
Product Type Test Tube Rack
Shelves 3 Tiers
Height (Metric) 75 mm
Array 6 x 6
Width (Metric) 158 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.